Learn More
Schisandrin C, MedChemExpress
Código de producto. 16429068
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
Tamaño de la unidad:
10 miligramos
16429068 10 mg 10 miligramos
1 options
Código de producto. 16429068 Proveedor Medchem Express N.º de proveedor HYN0690
MedChemExpress Schisandrin C (Schizandrin-C) is a phytochemical lignan isolated from Schizandra chinensis. Schisandrin C has diverse biological activities, including anticancer, anti-inflammatory and antioxidant effects. Schisandrin C can be used for cancer, alzheimer’s disease, and liver diseases research. Schisandrin C induces cell apoptosis.
Schisandrin C (Schizandrin-C) is a phytochemical lignan isolated from Schizandra chinensis. Schisandrin C has diverse biological activities, including anticancer, anti-inflammatory and antioxidant effects. Schisandrin C can be used for cancer, alzheimer’s disease, and liver diseases research. Schisandrin C induces cell apoptosis.
Especificaciones
Nombre del producto químico o material Schisandrin C
CAS 61301-33-5
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C22H24O6
Cantidad 10 mg
Sinónimo Schizandrin-C Wuweizisu-C
Información de solubilidad DMSO : 8.33 mg/mL (21.67 mM; Need ultrasonic)
SMILES C[C@H]([C@H](C)C1)CC2=C(C3=C1C=C4OCOC4=C3OC)C(OC)=C5OCOC5=C2
Peso molecular (g/mol) 384.42
Formula Weight (peso de la fórmula) 384.42
Porcentaje de pureza 99.85%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-Kinase/protease