Learn More
SCH 336, Tocris Bioscience™
Highly potent and selective CB2 inverse agonist
Marca: Tocris Bioscience 5815/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
SCH 336 is a highly potent and selective CB2 receptor inverse agonist (Ki = 1.8 nM, EC50 = 2 nM). Displays 100-fold selectivity for CB2 receptors over CB1. Increases forskolin stimulated cAMP accumulation in CHO cells expressing human CB2 receptors. Inhibits leukocyte migration in a murine model of delayed-type hypersensitivity and inhibits antigen-induced lung eosinophilia in a mouse allergy model.Especificaciones
| N-[(1S)-1-[4-[[4-Methoxy-2-[(4-methoxyphenyl)sulfonyl]phenyl]sulfonyl]phenyl]ethyl]methanesulfonamide | |
| 447459-51-0 | |
| >98% | |
| 50 mg |