Learn More
Tocris Bioscience™ sBADA
Descripción
sBADA is a green sulfonated BODIPY-FL 3-amino-D-alanine (sBADA) or fluorescent D-amino acid (FFDA). Labels peptidoglycans in live bacteria. Sulfonated form of BADA with increased hydrophilicity and thermostability. Excitation/emission λ ∽490/510 nm.
Especificaciones
Especificaciones
| Nombre del producto químico o material | (R)-2-Amino-3-(3-(5,5-difluoro-7,9-dimethyl-2-sulfo-5H-4λ4,5λ4-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-3-yl)propanamido)propanoic acid |
| CAS | 2376838-11-6 |
| Cantidad | 1 mg |
| Objetivo | Fluorescent Probes for Imaging Bacteria |
| Fórmula molecular | C17H21BF2N4O6S |
| Pureza | 0.95 |