Learn More
Tocris Bioscience™ SB 706504
Descripción
SB 706504 is a p38 MAPK inhibitor. Inhibits LPS-induced transcription of a range of chemokines and cytokines in chronic obstructive pulmonary disease (COPD) monocyte derived macrophages (MDMs). Also inhibits LPS-induced protein expression of IL-6, IL-10, TNFα and γ-inducible protein 10 in COPD MDMs. Effects on suppression of LPS-induced gene and protein expression are enhanced in combination with dexamethasone (Cat. No. 1126).
Especificaciones
Especificaciones
| Nombre del producto químico o material | N-Cyano-N'-[2-[[8-(2,6-difluorophenyl)-4-(4-fluoro-2-methylphenyl)-7,8-dihydro-7-oxopyrido[2,3-d]pyrimidin-2-yl]amino]ethyl]guanidine |
| CAS | 911110-38-8 |
| Cantidad | 10 mg |
| Fórmula molecular | C24H19F3N8O |
| Formula Weight (peso de la fórmula) | 492.46 |
| Pureza | 98% |