Learn More
Tocris Bioscience™ SB 332235
Descripción
SB 332235 is a potent CXCR2 antagonist (IC50 = 7.7 nM). Exhibits 285-fold selectivity for CXCR2 over CXCR1. Reduces leukocyte numbers in synovial fluid in acute and chronic rabbit arthritis models. Also reduces synovial fluid eicosanoid and cytokine levels in a rabbit chronic arthritis model. Inhibits proliferation of AML cell lines in vitro.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 6-Chloro-3-[[[(2,3-dichlorophenyl)amino]carbonyl]amino]-2-hydroxybenzenesulfonamide |
| CAS | 276702-15-9 |
| Cantidad | 10 mg |
| Objetivo | Chemokine Receptor Antagonists |
| Fórmula molecular | C13H10Cl3N3O4S |
| Formula Weight (peso de la fórmula) | 410.66 |
| Pureza | >98% |