Learn More
Tocris Bioscience™ Sazetidine A dihydrochloride
Descripción
Subtype-selective α4β2 nicotinic acetylcholine receptor ligand (Ki values are 0.26 and 54 nM at α4β2 and α3β4 receptors respectively). May act as a silent desensitizer or as an agonist, depending on subunit stoichiometry (EC50 = 1.1 nM for nAChR-stimulated dopamine release). Exhibits analgesic activity in vivo and significantly reduces nicotine self-administration in an experimental rat model.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Sazetidine A dihydrochloride |
| CAS | 2455450-63-0 |
| Cantidad | 10 mg |
| Fórmula molecular | C15H22Cl2N2O2 |
| Formula Weight (peso de la fórmula) | 333.25 |
| Pureza | >98% |
| Peso molecular | 333.25 |