Learn More
Saxagliptin hydrochloride, Tocris Bioscience™
GREENER_CHOICE
DPPIV/CD26 Inhibitors
Marca:  Tocris Bioscience 6507/50
Encompass
Código de producto. 15907970
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos »
Descripción
Descripción
Saxagliptin hydrochloride is a high affinity DPP-IV inhibitor (Ki = 0.6 nM). Inhibits plasma DPP-IV (87%). Elicits enhanced glucose clearance in Zuckerfa/fa rats.
Especificaciones
Especificaciones
709031-78-7
(1S,3S,5S)-2-[(2S)-Amino(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)acetyl]-2-azabicyclo[3.1.0]hexane-3-carbonitrile hydrochloride