missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SARS-CoV-2 Spike Protein S1/S2 Polyclonal Antibody

Código de producto. 17191453 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
0.10 miligramo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17191453 100 μg 0.10 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17191453 Proveedor Invitrogen™ N.º de proveedor PA5112048

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

The immunogen is located within the last 50 amino acids of SARS-CoV-2 (COVID-19, 2019-nCoV) Spike protein. Predicted reactivity based on immunogen sequence: SARS-CoV Spike proteins: (100%).

SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold symptoms to severe effects including death and respiratory distress. The CoV Spike (S) protein plays the most important roles in viral attachment, fusion and entry, and serves as a target for development of antibodies, entry inhibitors and vaccines. Of note is the Spike protein receptor-binding domain (RBD, S-RBD) in SARS-CoV-2 Spike protein which binds strongly to human and bat angiotensin-converting enzyme 2 (ACE2) receptors. SARS-CoV-2 RBD exhibits significantly higher binding affinity to ACE2 receptor than SARS-CoV RBD in comparison. S1 contains a receptor-binding domain (RBD) that can specifically bind to angiotensin-converting enzyme 2 (ACE2), the receptor on target cells. SARS-CoV-2 S is 1273 amino acids long and consists of a signal peptide (amino acids 1-13) located at the N-terminus, the S1 subunit (14-685 residues), and the S2 subunit (686-1273 residues); the last two regions are responsible for receptor binding and membrane fusion, respectively. The S-ECD is the extracellular domain of the Spike protein.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno SARS-CoV-2 Spike Protein S1/S2
Aplicaciones ELISA, Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.02% sodium azide
Alias de gen Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2
Especie del huésped Rabbit
Inmunógeno peptide corresponding to 20 amino acids near the carboxy terminus of SARS-CoV-2(COVID-19, 2019-nCoV) Spike glycoprotein.
Método de purificación Antigen affinity chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana Virus
Contenido y almacenamiento 4°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.