Learn More
SAR407899 hydrochloride, MedChemExpress
Código de producto. 16466876
Encompass_Preferred
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Cantidad:
5 mg
10 mg
50 mg
100 mg
missing translation for 'unitSize'
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
16466876 100 mg 100 miligramos
16439297 5 mg 5 miligramos
16416748 10 mg 10 miligramos
16412827 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 16466876 missing translation for 'mfr' Medchem Express missing translation for 'supplierNo' HY15687
This item is not returnable. View return policy
MedChemExpress SAR407899 hydrochloride is a selective, potent and ATP-competitive ROCK inhibitor, with an IC50 of 135 nM for ROCK-2, and Kis of 36 nM and 41 nM for human and rat ROCK-2, respectively.
SAR407899 hydrochloride is a selective, potent and ATP-competitive ROCK inhibitor, with an IC50 of 135 nM for ROCK-2, and Kis of 36 nM and 41 nM for human and rat ROCK-2, respectively.
Specifications
Nombre del producto químico o material SAR407899 hydrochloride
CAS 923262-96-8
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C14H17ClN2O2
Cantidad 100 mg
Información de solubilidad H2O : 50 mg/mL (178.09 mM; Need ultrasonic) ∣DMSO : 25 mg/mL (89.05 mM; Need ultrasonic)
SMILES O=C(NC=C1)C(C1=C2)=CC=C2OC3CCNCC3.[H]Cl
Peso molecular (g/mol) 280.75
Formula Weight (peso de la fórmula) 280.75
Porcentaje de pureza 98.0%
Grado Research
Forma física Powder
Para utilizar con (aplicación) Metabolism-protein/nucleotide metabolism