Learn More
Salvianolic acid C, MedChemExpress
Código de producto. 16471378
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
Tamaño de la unidad:
10 miligramos
25 miligramos
5 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16471378 5 mg 5 miligramos
16453856 25 mg 25 miligramos
16448948 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16471378 Proveedor Medchem Express N.º de proveedor HYN0319
MedChemExpress Salvianolic acid C is a noncompetitive Cytochrome P4502C8 (CYP2C8) inhibitor and a moderate mixed inhibitor of Cytochrome P45022J2 (CYP2J2), with Kis of 4.82 μM and 5.75 μM for CYP2C8 and CYP2J2, respectively.
Salvianolic acid C is a noncompetitive Cytochrome P4502C8 (CYP2C8) inhibitor and a moderate mixed inhibitor of Cytochrome P45022J2 (CYP2J2), with Kis of 4.82 μM and 5.75 μM for CYP2C8 and CYP2J2, respectively.
Especificaciones
Nombre del producto químico o material Salvianolic acid C
CAS 115841-09-3
Color Yellow
Notas de grado de pureza Research
Fórmula molecular C26H20O10
Cantidad 5 mg
Información de solubilidad DMSO : 50 mg/mL (101.54 mM; Need ultrasonic)
SMILES OC1=C(O)C=CC(C[C@H](C(O)=O)OC(/C=C/C2=CC=C(O)C3=C2C=C(C4=CC(O)=C(O)C=C4)O3)=O)=C1
Peso molecular (g/mol) 492.43
Formula Weight (peso de la fórmula) 492.43
Porcentaje de pureza 98.51%
Grado Research
Forma física Powder
Para utilizar con (aplicación) Cancer-programmed cell death