Learn More
Salirasib, MedChemExpress
Código de producto. 16496536
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16496536 100 mg 100 miligramos
16426158 10 mg 10 miligramos
16402297 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16496536 Proveedor Medchem Express N.º de proveedor HY14754
MedChemExpress Salirasib is a Ras inhibitor that inhibits specifically both oncogenically activated Ras and growth factor receptor-mediated Ras activation, resulting in the inhibition of Ras-dependent tumor growth.
Salirasib is a Ras inhibitor that inhibits specifically both oncogenically activated Ras and growth factor receptor-mediated Ras activation, resulting in the inhibition of Ras-dependent tumor growth.
Especificaciones
Nombre del producto químico o material Salirasib
Forma física Solid
CAS 162520-00-5
Color Off-White
Cantidad 100 mg
Notas de grado de pureza Research
Fórmula molecular C22H30O2S
Para utilizar con (aplicación) Cancer-Kinase/protease
Sinónimo S-Farnesylthiosalicylic acid Farnesyl Thiosalicylic Acid FTS
Información de solubilidad DMSO : ≥ 50 mg/mL (139.45 mM)
SMILES O=C(O)C1=CC=CC=C1SC/C=C(C)/CC/C=C(C)/CC/C=C(C)/C
Peso molecular (g/mol) 358.54
Formula Weight (peso de la fórmula) 358.54
Porcentaje de pureza 97.98%