Learn More
Tocris Bioscience™ SAK 3
Descripción
SAK 3 is a potent CaV3.1 and CaV3.3 activator. Enhances CaV3.1 and CaV3.3, but not CaV3.2 activity at concentrations of 0.01 - 10 M in neuro2A cells in vitro. Elevates ACh levels in hippocampus. Rescues impaired memory-related behaviors, promotes adult neurogenesis and displays antidepressant-like effects in OBX mice. Orally bioavailable.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Ethyl 8′-methyl-2′,4-dioxo-2-(piperidin-1-yl)-2′H-spiro[cyclopent[2]ene-1,3′-imidazo[1,2-a]pyridine]-3-carboxylate |
| CAS | 1256269-87-0 |
| Cantidad | 5 mg |
| Objetivo | Cav3.x Channel Activators |
| Fórmula molecular | C20H23N3O4 |
| Pureza | 98% |