missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SAFB Polyclonal Antibody
GREENER_CHOICE

Código de producto. 13208958
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
13208958 100 μL 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 13208958 Proveedor Invitrogen™ N.º de proveedor PA521356

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Recommended positive controls: A431, H1299, HeLa, HepG2. Predicted reactivity: Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a DNA-binding protein which has high specificity for scaffold or matrix attachment region DNA elements (S/MAR DNA). This protein is thought to be involved in attaching the base of chromatin loops to the nuclear matrix but there is conflicting evidence as to whether this protein is a component of chromatin or a nuclear matrix protein. Scaffold attachment factors are a specific subset of nuclear matrix proteins (NMP) that specifically bind to S/MAR. The encoded protein is thought to serve as a molecular base to assemble a 'transcriptosome complex' in the vicinity of actively transcribed genes. It is involved in the regulation of heat shock protein 27 transcription, can act as an estrogen receptor co-repressor and is a candidate for breast tumorigenesis. This gene is arranged head-to-head with a similar gene whose product has the same functions. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno SAFB
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
génica SAFB
N.º de referencia del gen D3YXK2, O88453, Q15424
Alias de gen 3110021E02Rik; 5330423C17Rik; AU018122; D18386; DKFZp779C1727; E130307D12; glutathione S-transferase fusion protein; HAP; heat-shock protein (HSP27) estrogen response element and TATA box-binding protein; HET; Hsp27 ERE-TATA binding protein; HSP27 ERE-TATA-binding protein; HSP27 estrogen response element-TATA box-binding protein; SAB-B1; Safb; SAF-B; Safb1; SAF-B1; scaffold attachment factor B; scaffold attachment factor B1
Símbolos de los genes SAFB
Especie del huésped Rabbit
Inmunógeno Recombinant protein encompassing a sequence within the center region of human SAFB. The exact sequence is proprietary.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 224903, 6294, 64196
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.