Learn More
Tocris Bioscience™ SA-15-P
Descripción
SA-15-P is a dual inhibitor of the interaction of lymphocyte activation gene 3 (LAG3) with both the major histocompatibility complex class II (MHCII) and fibrinogen-like protein 1 (FGL1) (IC50 values from TR-FRET assay are 4.2 μM and 6.5 μM respectively).
Especificaciones
Especificaciones
| Nombre del producto químico o material | N-[4-[2-[2,5-Dimethyl-1-(phenylmethyl)-1H-pyrrol-3-yl]-2-oxoethoxy]phenyl]-2-furancarboxamide |
| CAS | 852846-00-5 |
| Cantidad | 50 mg |
| Objetivo | LAG-3 |
| Fórmula molecular | C26H24N2O4 |
| Pureza | 0.98 |