Learn More
Brady™ S3100 Sign and Label Printer
Código de producto. 31842972
Change view
Click to view available options
Keyboard Type:
AZERTY
QWERTY
QWERTZ
Tamaño de la unidad:
1 set
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Keyboard Type unitSize
31842972 QWERTY 1 set
31842975 AZERTY 1 set
31842971 QWERTZ 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 31842972 Proveedor Brady™ N.º de proveedor 149126
The easy-to-use Brady Printer S3100 sign and label printer is designed to meet a variety of your facility identification needs, helping you save time so you can get back to work.
  • Create the labels you need, when you need them without the wait with do-it-yourself label creation
  • Ideal for multi-user areas in your facility, anyone can simply walk up, create labels and print right on the spot
  • The built-in keyboard and touchscreen interface make label creation a breeze
  • Label creation is even faster with the quick setup and print speeds that come with print smart technology
  • Prints on a variety of material options so you can create signs and labels for a variety of applications
  • Ships with free Brady Workstation Basic Label Design software suite, which will enable you to immediately design professional labels.
Especificaciones
Tipo de producto Printer
Color Blue, Gray
Tipo de pantalla Color LCD Touchscreen
Para utilizar con (equipo) B30 Series Thermal Transfer Tapes and Ribbons, B30 Series Thermal Transfer Tapes
Material Plastic
Print Technology Thermal Transfer
Longitud (imperial) 10.25 in.
Anchura (imperial) 12 in.
Longitud (métrico) 260.35 mm
Anchura (métrico) 304.80 mm
Altura (imperial) 9.5 in.
Altura (métrico) 241.30 mm
Dimensiones (Largo x Ancho x Alto) 260.35 x 304.80 x 241.30 mm
Esterilizables en autoclave Not Autoclavable
Esterilidad Non-sterile
Capacidad de color Monocolour
Tamaño de la pantalla 155.44 x 91.95 mm
Incluye S3100 Printer, Power Cord, USB Cable, Cleaning Kit, Cutter Cleaning Tool, Quick Start Guide, Brady Workstation Basic Suite, Stylus and Mouse
Idiomas Bulgarian, Chinese (Simplified), Chinese (Traditional), Croatian, Czech, Danish, Dutch, English(US), Estonian, Finnish, French, German, Hungarian, Italian, Korean, Norwegian, Polish, Portuguese, Romanian, Russian, Slovak, Slovenian, Spanish
Memoria 256 MB
Modelo S3100-QY-EU
Velocidad de impresión 76.00 mm/sec.
Resolución 300 dpi
Tipos de conector Ethernet, USB
Peso (imperial) 20 lb.
Peso (métrico) 9.07 kg
Certificaciones/conformidad UL, cUL, CE, WEEE, RoHS, RCM (Australia/New Zealand), EAC (Russia, Belarus, and Kazakhstan), China: CCC-RoHS
Para utilizar con (aplicación) Asset tracking, Cold temperature labelling, Component and equipment labelling, Electrical labelling, Exit and egress marking, Facility identification, Floor marking, General identification, Hazard communication identification, Lean and 5S labelling, Lockout tagout, Pipe and valve marking, Safety identification,Temperature indicating labelling
Cantidad 1 Ea.
Garantía 1 Year
Tensión 110/240 V
Keyboard Type QWERTY
Alternate Part Number S3100-QY-EU
Tipo de tapón AC Adaptor