Learn More
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 97%, Thermo Scientific™
Marca: Thermo Scientific Alfa Aesar B23872.01
| Cantidad | 0.25g |
|---|
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Especificaciones
| (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl | |
| C44H32P2 | |
| 0.25g | |
| Air Sensitive | |
| binap, r-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl, 2,2'-bis diphenylphosphino-1,1'-binaphthyl, s-binap, s---binap, s---2,2'-bis diphenylphosphino-1,1'-binaphthyl, r-+-binap, r-binap, rac-binap, 2,2'-bis diphenylphosphino-1,1'-binaphthalene | |
| −230° (c=0.3 in Toluene) | |
| [1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane | |
| 634876 | |
| 97% |
| 76189-56-5 | |
| MFCD00010805 | |
| 5321444 | |
| 14,1223 | |
| MUALRAIOVNYAIW-UHFFFAOYSA-N | |
| C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8 | |
| 622.688 | |
| 622.69 |