missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RSU1 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15743191 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15743191 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15743191 Proveedor Invitrogen™ N.º de proveedor PA555270

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Immunogen sequence: DLTYNNLSEN SLPGNFFYLT TLRALYLSDN DFEILPPDIG KLTKLQILSL RDNDLISLPK EIGELTQLKE LHIQGNRLTV LPPELGNLDL TGQKQVFKAE NNPWVTPIAD QFQLGVSHVF EYIRSETYKY LYGRHMQANP EPPKKN Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 96%.

RSU1 is a protein that is involved in the Ras signal transduction pathway, growth inhibition, and nerve-growth factor induced differentiation processes, as determined in mouse and human cell line studies. In mouse, the protein was initially isolated based on its ability to inhibit v-Ras transformation. This gene encodes a protein that is involved in the Ras signal transduction pathway, growth inhibition, and nerve-growth factor induced differentiation processes, as determined in mouse and human cell line studies. In mouse, the encoded protein was initially isolated based on its ability to inhibit v-Ras transformation. Multiple alternatively spliced transcript variants for this gene have been reported; one of these variants was found only in glioma tumors.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno RSU1
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 0.10 mg/mL
Conjugado Unconjugated
Formulación PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
génica RSU1
N.º de referencia del gen Q01730, Q15404
Alias de gen cubilin; ras suppressor protein 1; etID12695; FLJ31034; LOC553276 protein; ras suppressor protein 1; ras suppressor protein 1 isoform 1; ras suppressor protein 1 variant 1; ras suppressor protein 1 variant 2; ras suppressor protein 1 variant 3; ras suppressor protein 1 variant 5; Ras suppressor protein 1-like protein; RSP1; RSP-1; rsu1; Rsu-1; RsuI; si:dz63m2.1; Unknown (protein for MGC:126965)
Símbolos de los genes RSU1
Especie del huésped Rabbit
Inmunógeno Recombinant protein corresponding to Human RSU1. Recombinant protein control fragment (Product #RP-89461).
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 20163, 6251, 680419
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.