Learn More
STEMCELL Technologies RSC-133
Descripción
- Reprogramming stimulating compound (RSC)-133 is an indole derivative that specifically inhibits DNA methyltransferase DNMT1 (Lee et al.)
- Increases kinetics and efficiency of reprogramming human foreskin fibroblasts to induced pluripotent stem cells, when used with reprogramming factors OCT4, SOX2, KLF4, and c-MYC (Lee et al.)
- Supports the maintenance of human pluripotent stem cells in unconditioned medium (Lee et al.)
Especificaciones
Especificaciones
| CAS | 1418131-46-0 |
| Cantidad | 10 mg |
| Fórmula molecular | C18H15N3O2 |
| Pureza | ≥95% |