Learn More
Tocris Bioscience™ (RS)-3,5-DHPG
GREENER_CHOICE
Selective group I mGlu agonist
Marca:  Tocris Bioscience™ 0342/10
Encompass
Código de producto. 13417573
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos »
Descripción
Descripción
(RS)-3,5-DHPG is a selective group I metabotropic glutamate receptor agonist which activates both mGlu1 and mGlu5. Also reported to be an antagonist at metabotropic glutamate receptors linked to phospholipase D.
S-enantiomer also available.
Especificaciones
Especificaciones
(RS)-3,5-Dihydroxyphenylglycine
>99%
Soluble to 10mM in water
19641-83-9
10 mg
Certificados
Promociones
Promociones
Título del producto
Seleccione un problema
For Research Use Only