Learn More
RR6, MedChemExpress
Código de producto. 16481749
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
1 mg
5 mg
10 mg
25 mg
50 mg
Tamaño de la unidad:
1 miligramo
10 miligramos
25 miligramos
5 miligramos
50 miligramos
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
16481749 1 mg 1 miligramo
16453647 50 mg 50 miligramos
16453616 25 mg 25 miligramos
16437638 10 mg 10 miligramos
16410108 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Código de producto. 16481749 Proveedor Medchem Express N.º de proveedor HY18780
MedChemExpress RR6 is a potent, selective, reversible, competitive and orally active vanin inhibitor with an IC50 of 540 nM for recombinant vanin-1. RR6 also potently inhibits human, bovine and rat serum pantetheinase with IC50 values of 40 nM, 41 nM and 87 nM, respectively.
RR6 is a potent, selective, reversible, competitive and orally active vanin inhibitor with an IC50 of 540 nM for recombinant vanin-1. RR6 also potently inhibits human, bovine and rat serum pantetheinase with IC50 values of 40 nM, 41 nM and 87 nM, respectively.
Especificaciones
Nombre del producto químico o material RR6
Forma física Solid
CAS 1351758-37-6
Color Off-White
Cantidad 1 mg
Notas de grado de pureza Research
Fórmula molecular C16H23NO4
Para utilizar con (aplicación) Cancer-programmed cell death
Información de solubilidad DMSO : ≥ 100 mg/mL (340.88 mM)
SMILES CC(C)(CO)[C@@H](O)C(NCCC(CC1=CC=CC=C1)=O)=O
Peso molecular (g/mol) 293.36
Formula Weight (peso de la fórmula) 293.36
Porcentaje de pureza 99.93%