Learn More
Rostafuroxin, MedChemExpress
Código de producto. 16471297
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16471297 50 mg 50 miligramos
16405068 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16471297 Proveedor Medchem Express N.º de proveedor HY12283
MedChemExpress Rostafuroxin (PST 2238), a digitoxigenin derivative, is an orally active and potent Na+,K+-ATPase (ATP1A1) antognist. Rostafuroxin binds specifically to the ATP1A1 extracellular domain and blocks respiratory syncytial virus (RSV)-triggered EGFR Tyr845 phosphorylation. Rostafuroxin has antihypertensive and anti-RSV activity.
Rostafuroxin (PST 2238), a digitoxigenin derivative, is an orally active and potent Na+,K+-ATPase (ATP1A1) antognist. Rostafuroxin binds specifically to the ATP1A1 extracellular domain and blocks respiratory syncytial virus (RSV)-triggered EGFR Tyr845 phosphorylation. Rostafuroxin has antihypertensive and anti-RSV activity.
Especificaciones
Nombre del producto químico o material Rostafuroxin
CAS 156722-18-8
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C23H34O4
Cantidad 50 mg
Sinónimo PST 2238
Información de solubilidad DMSO : ≥ 50 mg/mL (133.51 mM)
SMILES O[C@]([C@@](CC[C@@]1([H])[C@@]2(CC[C@H](O)C1)C)([H])[C@]2([H])CC3)(CC[C@@]4(C5=COC=C5)O)[C@]34C
Peso molecular (g/mol) 374.51
Formula Weight (peso de la fórmula) 374.51
Porcentaje de pureza 98.07%
Grado Research
Forma física Solid
Para utilizar con (aplicación) COVID-19-immunoregulation