Learn More
Tocris Bioscience™ Rostafuroxin
Descripción
Rostafuroxin is a Na+/K+ ATPase modulator. Inhibits the Normalizes renal Na+/K+ ATPase pump function in mutant adducin- and endogenous ouabain-dependent forms of hypertension. Reduces blood pressure in Milan hypertensive rats. Controls mania-like behaviors in Myshkin mice having a Na+/K+ ATPase α3 subunit mutation. Antagonizes the effects of Ouabain (Cat.No. 1076) on myogenic tone and Ca2+ signaling in rat hippocampal neurons.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 3β,5β,14β)-21,23-Epoxy-24-norchola-20,22-diene-3,14,17-triol |
| CAS | 156722-18-8 |
| Sinónimo | PST 2238 |
| Cantidad | 50 mg |
| Objetivo | Na+/K+ ATPase Modulators |
| Fórmula molecular | C23H34O4 |
| Formula Weight (peso de la fórmula) | 374.51 |
| Pureza | >98% |