Learn More
Tocris Bioscience™ Roscovitine
Descripción
Cyclin-dependent kinase (cdk) inhibitor. Displays sub-micromolar inhibition of cdk1, cdk2, cdk5, cdk7 and cdk9 (reported IC50 values range from 0.16 - 0.28 μM for cdk5 to 0.8 μM for cdk7). Selective for cdks over a wide range of related kinases including ERK1 and ERK2. Arrests L1210 cells in G1 phase. Inhibits phosphorylation of vimentin in vivo. Antimitotic.
Especificaciones
Especificaciones
| Nombre del producto químico o material | (2R)-2-[[9-(1-Methylethyl)-6-[(phenylmethyl)amino]-9H-purin-2-yl]amino]-1-butanol |
| CAS | 186692-46-6 |
| Sinónimo | roscovitine, rosco, 2-r-1-ethyl-2-hydroxyethylamino-6-benzylamino-9-isopropylpurine, unii-0es1c2kq94, insolution roscovitine, chembl14762, bmk1-e12 |
| Cantidad | 50 mg |
| Solubilidad | Soluble to 100mM in DMSO and to 100mM in ethanol |
| Fórmula molecular | C19H26N6O |
| Formula Weight (peso de la fórmula) | 354.45 |