missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ROS1 Monoclonal Antibody (OTI1A1)
GREENER_CHOICE

Código de producto. 15936354
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15936354 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15936354 Proveedor Invitrogen™ N.º de proveedor MA526760

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

ROS1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

ROS1 is a tyrosine kinase that functions as a growth or differentiation factor receptor. Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The tyrosine kinase (TK) group is mainly involved in the regulation of cell-cell interactions such as differentiation, adhesion, motility and death. There are currently about 90 TK genes sequenced, 58 are of receptor protein TK (e.g. EGFR, EPH, FGFR, PDGFR, TRK, and VEGFR families), and 32 of cytosolic TK (e.g. ABL, FAK, JAK, and SRC families).
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno ROS1
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Monoclonal
Clon OTI1A1
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
génica ROS1
N.º de referencia del gen P08922, Q78DX7
Alias de gen c-ros; c-ros oncogene 1 , receptor tyrosine kinase; c-Ros receptor tyrosine kinase; c-ros-1; heart - derived c - ros - 1 proto - oncogene; MCF3; Proto-oncogene c-Ros; proto-oncogene c-Ros-1; Proto-oncogene tyrosine-protein kinase ROS; receptor tyrosine kinase c-ros oncogene 1; ROS; ROS proto-oncogene 1 , receptor tyrosine kinase; ROS proto-oncogene 1, receptor tyrosine kinase; Ros1; Ros-1; Ros1 proto-oncogene; ROS1C; RP1-179P9.1; transmembrane tyrosine-specific protein kinase; v-ros avian UR2 sarcoma virus oncogene homolog 1; v-ros UR2 sarcoma virus oncogene homolog 1
Símbolos de los genes ROS1
Especie del huésped Mouse
Inmunógeno Human recombinant protein fragment corresponding to amino acids 1882-2125 of ROS1 produced in E.coli.
Método de purificación Affinity Chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 19886, 6098
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2a
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.