Learn More
Rociletinib hydrobromide, MedChemExpress
Código de producto. 16486896
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16486896 100 mg 100 miligramos
16456778 10 mg 10 miligramos
16439327 5 mg 5 miligramos
16432857 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16486896 Proveedor Medchem Express N.º de proveedor HY15729A
MedChemExpress Rociletinib hydrobromide (CO-1686 hydrobromide) is an orally delivered kinase inhibitor that specifically targets the mutant forms of EGFR including T790M, and the Ki values for EGFRL858R/T790M and EGFRWT are 21.5 nM and 303.3 nM, respectively.
Rociletinib hydrobromide (CO-1686 hydrobromide) is an orally delivered kinase inhibitor that specifically targets the mutant forms of EGFR including T790M, and the Ki values for EGFRL858R/T790M and EGFRWT are 21.5 nM and 303.3 nM, respectively.
Especificaciones
Nombre del producto químico o material Rociletinib hydrobromide
CAS 1446700-26-0
Color Yellow
Notas de grado de pureza Research
Fórmula molecular C27H29BrF3N7O3
Cantidad 100 mg
Sinónimo CO-1686 hydrobromide AVL-301 hydrobromide CNX-419 hydrobromide
Información de solubilidad DMSO : ≥ 59 mg/mL (92.70 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C=CC(NC1=CC=CC(NC2=NC(NC3=CC=C(N4CCN(C(C)=O)CC4)C=C3OC)=NC=C2C(F)(F)F)=C1)=O.[H]Br
Peso molecular (g/mol) 636.46
Formula Weight (peso de la fórmula) 636.46
Porcentaje de pureza 98.04%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-Kinase/protease