Learn More
RNPA1000, MedChemExpress
Código de producto. 16498087
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16498087 5 mg 5 miligramos
16451567 50 mg 50 miligramos
16415378 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16498087 Proveedor Medchem Express N.º de proveedor HY12824
MedChemExpress RNPA1000, an antibiotic, is a potent RnpA inhibitor and inhibits RnpA-mediated cellular RNA degradation. RNPA1000 inhibits tRNA maturation with an IC50 of 175 μM. RNPA1000 displays broad-spectrum antimicrobial activities and inhibits staphylococcal and all Gram-positive bacterial pathogens activity.
RNPA1000, an antibiotic, is a potent RnpA inhibitor and inhibits RnpA-mediated cellular RNA degradation. RNPA1000 inhibits tRNA maturation with an IC50 of 175 μM. RNPA1000 displays broad-spectrum antimicrobial activities and inhibits staphylococcal and all Gram-positive bacterial pathogens activity.
Especificaciones
Nombre del producto químico o material RNPA1000
Forma física Solid
CAS 359600-10-5
Color Earth Yellow
Cantidad 5 mg
Notas de grado de pureza Research
Fórmula molecular C23H18BrN3O3
Para utilizar con (aplicación) Neuroscience-Neuromodulation
Información de solubilidad DMSO : 50 mg/mL (107.69 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=C(N2C(C)=C(/C=C(C#N)/C(NC3=CC=CC(Br)=C3)=O)C=C2C)C=C1
Peso molecular (g/mol) 464.31
Formula Weight (peso de la fórmula) 464.31
Porcentaje de pureza 98.0%