missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF19A Polyclonal Antibody

Descripción
Antibody detects endogenous levels of total RNF19A.
This gene encodes a member of the ring between ring fingers (RBR) protein family, and the encoded protein contains two RING-finger motifs and an in between RING fingers motif. This protein is an E3 ubiquitin ligase that is localized to Lewy bodies, and ubiquitylates synphilin-1, which is an interacting protein of alpha synuclein in neurons. The encoded protein may be involved in amyotrophic lateral sclerosis and Parkinson's disease. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Jul 2013].
Especificaciones
Especificaciones
| Antígeno | RNF19A |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | RNF19A |
| N.º de referencia del gen | P50636, Q9NV58 |
| Alias de gen | AA032313; Dorfin; Double ring-finger protein; E3 ubiquitin-protein ligase RNF19A; Gametogenesis-expressed protein GEG-154; Geg-154; p38; protein p38 interacting with transcription factor Sp1; ring finger protein (C3HC4 type) 19; ring finger protein 19; ring finger protein 19A; ring finger protein 19A, E3 ubiquitin protein ligase; ring finger protein 19A, RBR E3 ubiquitin protein ligase; ring-IBR-ring domain containing protein Dorfin; Rnf19; RNF19A; Ubce7ip2; UBCM4-interacting protein 117; ubiquitin conjugating enzyme 7 interacting protein 2; UIP117; XY body protein; XYbp |
| Símbolos de los genes | RNF19A |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?