Learn More
Tocris Bioscience™ RN 9893 hydrochloride
Descripción
RN 9893 hydrochloride is a potent and selective TRPV4 antagonist (IC50 values are 320, 420 and 660 nM for mouse, human and rat channels, respectively). Exhibits >15-fold selectivity for TRPV4 over TRPV1, TRPV3 and TRPM8, and a panel of 54 other common biological targets. Orally bioavailable.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N-[4-[[4-(1-Methylethyl)-1-piperazinyl]sulfonyl]phenyl]-2-nitro-4-(trifluoromethyl)benzamide hydrochloride |
| CAS | 2109450-40-8 |
| Cantidad | 50 mg |
| Objetivo | TRPV Antagonists |
| Fórmula molecular | C21H23F3N4O5S.HCl |
| Formula Weight (peso de la fórmula) | 536.95 |
| Pureza | >99% |