Learn More
Tocris Bioscience™ RN 1734
Descripción
RN 1734 is a selective TRPV4 antagonist (IC50 values are 2.3, 3.2 and 5.9 μM for hTRPV4, rTRPV4 and mTRPV4 receptors respectively). Displays selectivity for TRPV4 over other TRP channels (IC50 values are 2.3, >30, >30 and >100 μM for TRPV4, TRPV3, TRPM8 and TRPV1 respectively). Blocks TRPV4-mediated arteriole vasodilation and increases myogenic tone ex vivo.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 2,4-Dichloro-N-isopropyl-N-(2-isopropylaminoethyl)benzenesulfonamide |
| CAS | 946387-07-1 |
| Cantidad | 25 mg |
| Fórmula molecular | C14H22Cl2N2O2S |
| Formula Weight (peso de la fórmula) | Observed MW: 353.31 |