Learn More
Tocris Bioscience™ RKI 1447 dihydrochloride
GREENER_CHOICE
Potent and selective ROCK inhibitor; antitumor
Marca:  Tocris Bioscience™ 5061/10
Encompass
Código de producto. 15716615
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos »
Descripción
Descripción
RKI 1447 dihydrochloride is a potent and selective ROCK inhibitor (IC50 values are 6.2 and 14.5 nM for ROCK2 and ROCK1 respectively); suppresses phosphorylation of ROCK substrates MLC-2 and MYPT-1 in human cancer cells, but has no effect on the phosphorylation levels of Akt, MEK, and S6 kinase. Displays anti-invasive and antitumor activities in breast cancer cells.
Especificaciones
Especificaciones
>99%
1782109-09-4
10 mg
Certificados
Promociones
Promociones
Título del producto
Seleccione un problema
For Research Use Only