Learn More
Tocris Bioscience™ Ritonavir
HIV-1 and HIV-2 protease inhibitor
Marca: Tocris Bioscience™ 5856/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Ritonavir is a HIV-1 and HIV-2 protease inhibitor (EC50 values are 0.022-0.13 and 0.16 μM, respectively). Blocks the metabolism of protease inhibitors by liver enzyme cytochrome P450-3A4 (CYP3A4). Orally bioavailable. When used in combination with lopinavir, improves outcome in an animal model of MERS-CoV infection.Especificaciones
| 5-Thiazolylmethyl (3 S,4 S,6 S,9 S)-4-hydroxy-12-methyl-9-(1-methylethyl)-13-[2-(1-methylethyl)-4-thiazolyl]-8,11-dioxo-3,6-bis(phenylmethyl)-2,7,10,12-tetraazatridecanoate | |
| 155213-67-5 | |
| ABT-538, A-84538, Abbot 84538 | |
| 50 mg |