Learn More
Thermo Scientific™ Richard-Allan Scientific™ Cytoseal™™ 60 and 280, Cytoseal 60
Prevent cracking, fading and discoloration of stained specimens with Thermo Scientific™ Richard-Allan Scientific™ Cytoseal™ 60 and 280 mounting media.
Marca:  Thermo Scientific™ 8310-4
Código de producto. 10444920
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos »
Descripción
Descripción
Prevent cracking, fading and discoloration of stained specimens with Thermo Scientific™ Richard-Allan Scientific™ Cytoseal™ 60 and 280 mounting media.
  • Cytoseal 60 mountant is a low-viscosity medium that dries quickly and allows for rapid, even spreading which virtually eliminates air bubbles
  • Cytoseal 280 mountant is a high-viscosity medium for applications where minimal spreading is desired or applications without a cover glass
Ordering Information:
Packaged in an easy-to-use dispenser with a twist-open/twist-closed cap.
TRUSTED_SUSTAINABILITY
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones