missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RhoA/RhoC Monoclonal Antibody (1B3-4A10)

Descripción
MA1-123 detects GTPases RhoA and RhoC, but does not react with RhoB. MA1-123 has been successfully used in Western blot, immunofluorescence, IHC (P) and immunoprecipitation applications with human and mouse samples.
RhoA is described as a small GTPase protein that regulates the actin cytoskeleton in the formation of stress fibers. RhoA is also included in the related proteins of the Ras superfamily, which involves proteins in the regulation and timing of cell division.
Especificaciones
Especificaciones
| Antígeno | RhoA/RhoC |
| Aplicaciones | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | 1B3-4A10 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide |
| génica | RHOC |
| N.º de referencia del gen | P08134, P61586, P61589, Q62159, Q9QUI0 |
| Alias de gen | AI324259; Aplysia ras-related homolog 12; aplysia ras-related homolog 9 (RhoC); aplysia ras-related homolog A; aplysia ras-related homolog A1; aplysia ras-related homolog A2; ARH12; ARH9; ARHA; Arha1; Arha2; Arhc; h12; H9; oncogene RHO H12; oncogene RHO H9; plysia ras-related homolog A2; Ras family member A; ras homolog 9 (RhoC); ras homolog A1; ras homolog A2; ras homolog family member A; ras homolog family member C; ras homolog gene family, member A; ras homolog gene family, member A1; ras homolog gene family, member A2; ras homolog gene family, member C; RAS-related homolog 9; Rho; Rho cDNA clone 12; Rho cDNA clone 9; Rho family GTPase; RHO12; Rhoa; Rhoc; rhoC GTPase; RHOH12; RHOH9; rho-related GTP-binding protein RhoC; SIG-61; silica-induced gene 61 protein; small GTP binding protein RhoA; small GTP binding protein RhoC; transforming protein RhoA |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?