Learn More
STEMCELL Technologies Rho Kinase Inhibitor IV (Dihydrochloride)
Descripción
- Rho Kinase Inhibitor IV is a selective and potent inhibitor of Rho-associated coiled-coil containing protein kinase 2 (ROCK2; IC50 = 11.8 nM; Tamura et al.) It is a glycyl analog of Fasudil (Catalog #73662) with increased specificity for ROCK2 (Tamura)
- Rho Kinase Inhibitor IV is more potent than other ROCK inhibitors, including Y-27632 (Ki = 220 nM) and Fasudil (IC50 = 158 nM)
- It shows good specificity for ROCK2 compared to other kinases such as calcium/calmodulin-dependent kinase type II (IC50 = 2.57 μM), PKG (IC50 = 2.35 μM), Aurora A (IC50 = 3.26 μM), or PKA or PKC (IC50 ≥ 10 μM each)
- ROCK1 and ROCK2 act downstream of the G protein Rho to regulate actin-myosin turnover and dynamics, and play an important role in stem cell renewal, smooth muscle contraction, cell adhesion, and proliferation (Narumiya et al.; Olson; Watanabe et al.) This product is supplied as the dihydrochloride salt of the molecule
Especificaciones
Especificaciones
| CAS | 67-56-1 |
| Sinónimo | (S)-Glycyl-H-1152 |
| Cantidad | 1 mg |
| Fórmula molecular | C18H24N4O3S · 2HCl |
| Pureza | ≥98% |