Learn More
Rhapontin, TRC
Product Code. 30011233
Change view
Click to view available options
Cantidad:
25 mg
50 mg
Unit Size:
25 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30011233 50 mg 50 miligramos
30004922 25 mg 25 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011233 Supplier Toronto Research Chemicals Supplier No. TRCR31600050MG
This item is not returnable. View return policy
Moléculas orgánicas de alta pureza y estándares analíticos, implementados estratégicamente en todo el mundo para impulsar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 155-58-8
Fórmula molecular C21 H24 O9
Peso molecular (g/mol) 420.41
Sinónimo β-D-Glucopyranoside, 3-hydroxy-5-[(1E)-2-(3-hydroxy-4-methoxyphenyl)ethenyl]phenyl, Rhapontin (6CI,7CI,8CI), β-D-Glucopyranoside, 3-hydroxy-5-[2-(3-hydroxy-4-methoxyphenyl)ethenyl]phenyl, (E)-, 3,3',5-Trihydroxy-4'-methoxystilbene 3-β-D-glucopyranoside, 3-Hydroxy-5-[(1E)-2-(3-hydroxy-4-methoxyphenyl)ethenyl]phenyl β-D-glucopyranoside, 4'-Methoxy-3,3',5-stilbenetriol 3-β-D-glucopyranoside, NSC 43321, Ponticin, Rhaponticin, Rhaponticoside, Rhapontinum, trans-Rhapontin
Nombre IUPAC (2S,3R,4S,5S,6R)-2-[3-hidroxi-5-[(E)-2-(3-hidroxi-4-metoxifenil)etil]fenoxi]-6-(hidroximetil)oxano-3,4,5-triol
SMILES COc1ccc(\C=C\c2cc(O)cc(O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)c2)cc1O
Specifications
Nombre del producto químico o material Rhaponticin
Cantidad 50 mg
Fórmula InChI InChI=1S/C21H24O9/c1-28-16-5-4-11(8-15(16)24)2-3-12-6-13(23)9-14(7-12)29-21-20(27)19(26)18(25)17(10-22)30-21/h2-9,17-27H,10H2,1H3/b3-2+/t17-,18-,19+,20-,21-/m1/s1
Formula Weight (peso de la fórmula) 420.142