Learn More
Revexepride, MedChemExpress
Código de producto. 16452588
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
1 mg
5 mg
Tamaño de la unidad:
1 miligramo
5 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16452588 5 mg 5 miligramos
16423379 1 mg 1 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16452588 Proveedor Medchem Express N.º de proveedor HYU00373
MedChemExpress Revexepride is a highly selective 5-HT4 receptor agonist, and a potential inducer of CYP3A4 enzyme, used for the treatment of gastroesophageal reflux disease.
Revexepride is a highly selective 5-HT4 receptor agonist, and a potential inducer of CYP3A4 enzyme, used for the treatment of gastroesophageal reflux disease.
Especificaciones
Nombre del producto químico o material Revexepride
CAS 219984-49-3
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C21H32ClN3O4
Cantidad 5 mg
SMILES O=C(C1=C(OC(C)(C)C2)C2=C(N)C(Cl)=C1)NC[C@H]3[C@H](O)CN(CCCOC)CC3
Peso molecular (g/mol) 425.95
Formula Weight (peso de la fórmula) 425.95
Porcentaje de pureza 95.81%
Grado Research
Forma física Powder
Para utilizar con (aplicación) Neuroscience-Neuromodulation