Learn More
Resorcinol Bis (fosfato de difenilo), TRC
Product Code. 30006262
Change view
Click to view available options
Cantidad:
100 mg
250 mg
1 g
Unit Size:
1 g
100 miligramos
250 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30006262 1 g 1 g
30003906 250 mg 250 miligramos
30011085 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006262 Supplier Toronto Research Chemicals Supplier No. TRCR1453001G
This item is not returnable. View return policy
Moléculas orgánicas de alta pureza y estándares analíticos, implementados estratégicamente en todo el mundo para impulsar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 57583-54-7
Fórmula molecular C30 H24 O8 P2
Peso molecular (g/mol) 574.45
Sinónimo Phosphoric acid, P,P'-1,3-phenylene P,P,P',P'-tetraphenyl ester, Phosphoric acid, 1,3-phenylene tetraphenyl ester (9CI), 1,3-Phenylene bis(diphenyl phosphate), ADK Stab PFP, ADK Stab PFR, BPHPPO, CR 733S, Fyrolflex RDP, LDP 301, Mark PFK, Nonnen R 0111-10, PFR, PhireGuard RDP, Reofos RDP, Resorcinol bis(diphenyl phosphate), Resorcinol tetraphenyl diphosphate, Tetraphenyl m-phenylene bisphosphate, Tetraphenyl m-phenylene diphosphate, Tetraphenyl resorcinol diphosphate, WSFR-RDP, m-Phenylene bis(diphenyl phosphate)
Nombre IUPAC (3-difenoxifosforiloxifenilo) fosfato de difenilo
SMILES O=P(Oc1ccccc1)(Oc2ccccc2)Oc3cccc(OP(=O)(Oc4ccccc4)Oc5ccccc5)c3
Specifications
Nombre del producto químico o material Resorcinol Bis(diphenyl phosphate)
Cantidad 1 g
Fórmula InChI InChI=1S/C30H24O8P2/c31-39(33-25-14-5-1-6-15-25,34-26-16-7-2-8-17-26)37-29-22-13-23-30(24-29)38-40(32,35-27-18-9-3-10-19-27)36-28-20-11-4-12-21-28/h1-24H
Formula Weight (peso de la fórmula) 574.0946