Learn More
Resibufogenin, MedChemExpress
Código de producto. 16471518
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16471518 5 mg 5 miligramos
16454897 50 mg 50 miligramos
16429118 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16471518 Proveedor Medchem Express N.º de proveedor HYN0815
MedChemExpress Resibufogenin, a component of huachansu, has been shown to exhibit the anti-proliferative effect against cancer cells, and this may be attributed to the degradation of cyclin D1 caused by the activation of GSK-3β.
Resibufogenin, a component of huachansu, has been shown to exhibit the anti-proliferative effect against cancer cells, and this may be attributed to the degradation of cyclin D1 caused by the activation of GSK-3β.
Especificaciones
Nombre del producto químico o material Resibufogenin
CAS 465-39-4
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C24H32O4
Cantidad 5 mg
Sinónimo Bufogenin Recibufogenin
Información de solubilidad DMSO : 100 mg/mL (260.07 mM; Need ultrasonic)
SMILES C[C@]([C@@H](C(C=C1)=COC1=O)C2)(CC[C@@]3([H])[C@@]4([H])CC[C@@]5([H])[C@@]3(CC[C@H](O)C5)C)[C@@]64[C@@H]2O6
Peso molecular (g/mol) 384.51
Formula Weight (peso de la fórmula) 384.51
Porcentaje de pureza 99.33%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death