missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat SerpinF1 / PEDF Protein
A DNA sequence encoding the Rattus norvegicus (Rat) SerpinF1 / PEDF, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6675-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_808788.1 | |
SerpinF1 / PEDF Protein | |
Research Use Only | |
Serpinf1 | |
Recombinant Protein | |
E. coli | |
NVPDSSQDSP APDSTGEPVV EEDDPFFKAP VNKLAAAVSN FGYDLYRLRS GAVSTGNILL SPLSVATALS ALSLGAEQRT ESVIHRALYY DLINNPDIHS TYKELLASVT APEKNFKSAS RIVFERKLRV KSSFVAPLEK SYGTRPRILT GNPRIDLQEI NNWVQAQMKG KIARSTREMP SALSILLLGV AYFKGQWATK FDSRKTTLQD FHLDEDRTVR VPMMSDPKAI LRYGLDSDLN CKIAQLPLTG SMSIIFFLPL TVTQNLTMIE ESLTSEFVHD IDRELKTIQA VLTVPKLKLS YEGDVTNSLQ DMKLQSLFES PDFSKITGKP VKLTQVEHRA AFEWNEEGAG TSSNPDLQPV RLTFPLDYHL NRPFIFVLRD TDTGALLFIG RILDPSST | |
Greater than 90% as determined by SDS-PAGE. |
287526 | |
50 μg | |
Not determined. | |
Full Length | |
Rat | |
GST | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Rat SerpinF1 / PEDF Protein > 50μg
Spot an opportunity for improvement?Share a Content Correction