Learn More
RBPMS Polyclonal Antibody, Invitrogen™
Código de producto. 15711162 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
15711162 100 μL 100 microlitros
1 options
Código de producto. 15711162 Proveedor Invitrogen N.º de proveedor PA563383
Rabbit Polyclonal Antibody
Immunogen sequence: PSTPLPNTVP QFIAREPYEL TVPALYPSSP EV Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
This gene encodes a member of the RRM family of RNA-binding proteins. The RRM domain is between 80-100 amino acids in length and family members contain one to four copies of the domain. The RRM domain consists of two short stretches of conserved sequence called RNP1 and RNP2, as well as a few highly conserved hydrophobic residues. The protein encoded by this gene has a single, putative RRM domain in its N-terminus. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno RBPMS
Aplicaciones Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Clasificación Polyclonal
Concentración 0.1 mg/mL
Conjugado Unconjugated
Formulación PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
génica RBPMS
N.º de referencia del gen Q93062
Alias de gen HERMES, RBP-MS, RNA-binding protein with multiple splicing, heart and RRM expressed sequence
Símbolos de los genes RBPMS
Especie del huésped Rabbit
Inmunógeno Recombit protein corresponding to Human RBPMS
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 11030
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG