Learn More
CD4 Rat anti-Mouse, Clone: GK1.5, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Marca:  R&D Systems MAB554-100
Encompass
Código de producto. 15787504
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos »
Descripción
Descripción
CD4 Monoclonal specifically detects CD4 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Cell depletion, Costimulation of T Cells, Inhibition of T Cell Function, CyTOF-ready.
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones