Learn More
Rapamicina-d3 (contiene d0) grado técnico, TRC
Product Code. 30426647
Change view
Click to view available options
Cantidad:
10 mg
Unit Size:
10 miligramos
Product Code. Cantidad unitSize
30426647 10 mg 10 miligramos
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30426647 Supplier Toronto Research Chemicals Supplier No. TRCR12400210MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Moléculas orgánicas de alta pureza y estándares analíticos, entregados estratégicamente en todo el mundo para potenciar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material Sirolimus-d3
CAS 392711-19-2
Fórmula molecular C51 2H3 H76 N O13
Cantidad 10 mg
Sinónimo Rapamycin-d3, (-)-Rapamycin-d3
Fórmula InChI InChI=1S/C51H79NO13/c1-30-16-12-11-13-17-31(2)42(61-8)28-38-21-19-36(7)51(60,65-38)48(57)49(58)52-23-15-14-18-39(52)50(59)64-43(33(4)26-37-20-22-40(53)44(27-37)62-9)29-41(54)32(3)25-35(6)46(56)47(63-10)45(55)34(5)24-30/h11-13,16-17,25,30,32-34,36-40,42-44,46-47,53,56,60H,14-15,18-24,26-29H2,1-10H3/b13-11+,16-12+,31-17+,35-25+/t30-,32?,33-,34-,36-,37+,38+,39+,40-,42+,43+,44-,46-,47+,51-/m1/s1/i8D3
SMILES [2H]C([2H])([2H])O[C@H]1C[C@@H]2CC[C@@H](C)[C@@](O)(O2)C(=O)C(=O)N3CCCC[C@H]3C(=O)O[C@@H](CC(=O)[C@H](C)\C=C(/C)\[C@@H](O)[C@@H](OC)C(=O)[C@H](C)C[C@H](C)\C=C\C=C\C=C\1/C)[C@H](C)C[C@@H]4CC[C@@H](O)[C@@H](C4)OC
Peso molecular (g/mol) 917.19
Formula Weight (peso de la fórmula) 916.574