Learn More
Ranirestat, MedChemExpress
Código de producto. 16492547
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16492547 50 mg 50 miligramos
16456448 10 mg 10 miligramos
16429037 5 mg 5 miligramos
16416736 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16492547 Proveedor Medchem Express N.º de proveedor HY15314
MedChemExpress Ranirestat (AS-3201) potent and orally active aldose reductase (AR) inhibitor with IC50s of 11 nM and 15 nM for rat lens AR and recombinant human AR, respectively, and a Ki of 0.38 nM for recombinant human AR. Ranirestat has the potential for diabetic sensorimotor polyneuropathy treatment. Ranirestat also has a neuroprotective effect on diabetic retinas.
Ranirestat (AS-3201) potent and orally active aldose reductase (AR) inhibitor with IC50s of 11 nM and 15 nM for rat lens AR and recombinant human AR, respectively, and a Ki of 0.38 nM for recombinant human AR. Ranirestat has the potential for diabetic sensorimotor polyneuropathy treatment. Ranirestat also has a neuroprotective effect on diabetic retinas.
Especificaciones
Nombre del producto químico o material Ranirestat
CAS 147254-64-6
Color Off-White
Notas de grado de pureza Research
Fórmula molecular C17H11BrFN3O4
Cantidad 50 mg
Sinónimo AS-3201
Información de solubilidad DMSO : ≥ 50 mg/mL (118.99 mM)
SMILES O=C(N1CC2=CC=C(Br)C=C2F)C3=CC=CN3[C@]4(C(NC(C4)=O)=O)C1=O
Peso molecular (g/mol) 420.19
Formula Weight (peso de la fórmula) 420.19
Porcentaje de pureza 98.32%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Metabolism-sugar/lipid metabolism