Learn More
Metabolito de rameleón M-II-d3 (mezcla de R y S en posición hidroxi), TRC
Product Code. 30421488
Change view
Click to view available options
Cantidad:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad
30421488 10 mg
30416275 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30421488 Supplier Toronto Research Chemicals Supplier No. TRCR11006210MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Moléculas orgánicas de alta pureza y estándares analíticos, implementados estratégicamente en todo el mundo para impulsar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1246812-22-5
Fórmula molecular C16 D3 H18 N O3
Peso molecular (g/mol) 278.36
Sinónimo Ramelteon Metabolite M-II-d3 (RS-hydroxy)
Nombre IUPAC 3,3,3-trideuterio-2-hidroxi-N-[2-[(8S)-2,6,7,8-tetrahidro-1H-ciclopenta[e]benzofurano-8-il]etilpropanamida
SMILES [2H]C([2H])([2H])C(O)C(=O)NCC[C@@H]1CCc2ccc3OCCc3c12
Specifications
Nombre del producto químico o material 2-Hydroxy-N-[2-[(8S)-1,6,7,8-tetrahydro-2H-cyclopenta[e]benzofuran-8-yl]ethyl]propanamide-d3
Cantidad 10 mg
Fórmula InChI InChI=1S/C16H21NO3/c1-10(18)16(19)17-8-6-12-3-2-11-4-5-14-13(15(11)12)7-9-20-14/h4-5,10,12,18H,2-3,6-9H2,1H3,(H,17,19)/t10?,12-/m0/s1/i1D3
Formula Weight (peso de la fórmula) 278.171