Learn More
(Rac)-NSC305787 hydrochloride, MedChemExpress
Código de producto. 16497546
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16497546 100 mg 100 miligramos
16490108 5 mg 5 miligramos
16433657 50 mg 50 miligramos
16417648 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16497546 Proveedor Medchem Express N.º de proveedor HY18931A
MedChemExpress NSC305787 hydrochloride is an inhibitor of ezrin with a Kd of 5.85 μM, inhibits the phosphorylation of ezrin caused by PKCι with an IC50 of 8.3 μM, has antitumor activity.
NSC305787 hydrochloride is an inhibitor of ezrin with a Kd of 5.85 μM, inhibits the phosphorylation of ezrin caused by PKCι with an IC50 of 8.3 μM, has antitumor activity.
Especificaciones
Nombre del producto químico o material (Rac)-NSC305787 hydrochloride
Forma física Solid
CAS 53868-26-1
Color Blanco
Cantidad 100 mg
Notas de grado de pureza Research
Fórmula molecular C25H31Cl3N2O
Para utilizar con (aplicación) Cancer-Kinase/protease
Información de solubilidad DMSO : 8 mg/mL (16.60 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES OC(C1CCCCN1)C2=CC(C3(C[C@H](C4)C5)C[C@H]5C[C@H]4C3)=NC(C(Cl)=C6)=C2C=C6Cl.[H]Cl
Peso molecular (g/mol) 481.89
Formula Weight (peso de la fórmula) 481.89
Porcentaje de pureza 98.05%