Learn More
rac Mepindolol-d7, TRC
Product Code. 30473027
Change view
Click to view available options
Cantidad:
1 mg
10 mg
Unit Size:
1 miligramo
10 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30473027 1 mg 1 miligramo
30467337 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30473027 Supplier Toronto Research Chemicals Supplier No. TRCM2250271MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Moléculas orgánicas de alta pureza y estándares analíticos, implementados estratégicamente en todo el mundo para impulsar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1794970-97-0
Fórmula molecular C15 D7 H15 N2 O2
Peso molecular (g/mol) 269.39
Sinónimo 1-[(1-Methylethyl-d7)amino]-3-[(2-methyl-1H-indol-4-yl)oxy]-2-propanol, 1-[(Isopropyl-d7)amino]-3-[(2-methylindol-4-yl)oxy]-2-propanol, (±)-Mepindolol, -d7 Racemic Mepindolol-d7
Nombre IUPAC 1-(1,1,1,2,3,3,3,3-heptadeuteriopropan-2-ilamino)-3-[(2-metil-1H-indol-4-il)oxi]propano-2-ol
SMILES [2H]C([2H])([2H])C([2H])(NCC(O)COc1cccc2[nH]c(C)cc12)C([2H])([2H])[2H]
Specifications
Nombre del producto químico o material rac Mepindolol-d7
Cantidad 1 mg
Fórmula InChI InChI=1S/C15H22N2O2/c1-10(2)16-8-12(18)9-19-15-6-4-5-14-13(15)7-11(3)17-14/h4-7,10,12,16-18H,8-9H2,1-3H3/i1D3,2D3,10D
Formula Weight (peso de la fórmula) 269.212