Learn More
rac Desethyl Oxybutynin Hydrochlorur, TRC
Product Code. 30352206
Change view
Click to view available options
Cantidad:
10 mg
25 mg
50 mg
Unit Size:
10 miligramos
25 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30352206 25 mg 25 miligramos
30363492 50 mg 50 miligramos
30356009 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30352206 Supplier Toronto Research Chemicals Supplier No. TRCD28947525MG
This item is not returnable. View return policy
Moléculas orgánicas de alta pureza y estándares analíticos, implementados estratégicamente en todo el mundo para impulsar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 81039-77-2
Fórmula molecular C20 H27 N O3 . Cl H
Peso molecular (g/mol) 365.89
Sinónimo Desethyloxybutynin hydrochloride, Benzeneacetic acid, α-cyclohexyl-α-hydroxy-, 4-(ethylamino)-2-butyn-1-yl ester, hydrochloride (1:1) (ACI), Benzeneacetic acid, α-cyclohexyl-α-hydroxy-, 4-(ethylamino)-2-butynyl ester, hydrochloride (9CI), α-Cyclohexyl-α-hydroxybenzeneacetic acid 4-(ethylamino)-2-butyn-1-yl ester hydrochloride, 4-(Ethylamino)but-2-yn-1-yl 2-cyclohexyl-2-hydroxy-2-phenylacetate hydrochloride, N-Desethyloxybutynin hydrochloride, N-Desethyl Oxybutynin hydrochloride, rac Desethyl Oxybutynin hydrochloride
Nombre IUPAC 4-(etilamino)but-2-ynil 2-ciclohexil-2-hidroxi-2-fenilacetato; Cloruro
SMILES Cl.CCNCC#CCOC(=O)C(O)(C1CCCCC1)c2ccccc2
Specifications
Nombre del producto químico o material Desethyloxybutynin Hydrochloride
Cantidad 25 mg
Fórmula InChI InChI=1S/C20H27NO3.ClH/c1-2-21-15-9-10-16-24-19(22)20(23,17-11-5-3-6-12-17)18-13-7-4-8-14-18;/h3,5-6,11-12,18,21,23H,2,4,7-8,13-16H2,1H3;1H
Formula Weight (peso de la fórmula) 365.1758