Learn More
rac 1-oleoil-3-cloropropanodiol-d5, TRC
Product Code. 30414029
Change view
Click to view available options
Cantidad:
2.5 mg
Unit Size:
2.50 miligramo
Product Code. Cantidad unitSize
30414029 2.5 mg 2.50 miligramo
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30414029 Supplier Toronto Research Chemicals Supplier No. TRCO5268022.5MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Moléculas orgánicas de alta pureza y estándares analíticos, entregados estratégicamente en todo el mundo para potenciar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material rac 1-Oleoyl-3-chloropropanediol-d5
CAS 1246834-03-6
Fórmula molecular C21H34D5ClO3
Cantidad 2.5 mg
Sinónimo (9Z)-9-Octadecenoic Acid 3-Chloro-2-hydroxypropyl-d5 Ester, Oleic Acid 3-Chloro-2-hydroxypropyl-d5 Ester, 3-Chloro-1,2-propanediol-d5 1-Oleate, 3-Chloro-1,2-propanediol-d5 1-Monooleate
Fórmula InChI InChI=1S/C21H39ClO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-19-20(23)18-22/h9-10,20,23H,2-8,11-19H2,1H3/b10-9-/i18D2,19D2,20D
SMILES O=C(CCCCCCC/C=C\CCCCCCCC)OC([2H])([2H])C(O)([2H])C([2H])([2H])Cl
Nombre IUPAC Oleato de 3-cloro-2-hidroxipropil-1,1,2,3,3-d5
Peso molecular (g/mol) 380.02
Formula Weight (peso de la fórmula) 379.29