missing translation for 'onlineSavingsMsg'
Learn More

FANCD2 (phospho S222), Rabbit anti-Human, Polyclonal Antibody, Abnova™

Código de producto. 16029916
Cambiar vista
Click to view available options
Cantidad:
200 μL
Tamaño de la unidad:
200 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16029916 200 μL 200 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 16029916 Proveedor Abnova N.º de proveedor PAB12530.200uL

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit polyclonal antibody raised against synthetic phosphopeptide of FANCD2.

The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group D2. This protein is monoubiquinated in response to DNA damage, resulting in its localization to nuclear foci with other proteins (BRCA1 AND BRCA2) involved in homology-directed DNA repair. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq

Especificaciones

Antígeno FANCD2
Aplicaciones Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Descripción Rabbit polyclonal antibody raised against synthetic phosphopeptide of FANCD2.
Dilución The optimal working dilution should be determined by the end user.
Formulación No additive
génica FANCD2
Alias de gen DKFZp762A223/FA-D2/FA4/FACD/FAD/FAD2/FANCD/FLJ23826
Símbolos de los genes FANCD2
Especie del huésped Rabbit
Inmunógeno Synthetic phosphopeptide corresponding to residues surrounding S222 of human FANCD2.
Cantidad 200 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 2177
Especificidad de la prueba This antibody is specific to the phosphorylated form of FANCD2.
Especies diana Human
Contenido y almacenamiento Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Formulario Liquid
Mostrar más Mostrar menos

For Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.