Learn More
CDK2 (phospho T160), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Descripción
The protein encoded by this gene is a member of the Ser/Thr protein kinase family. This protein kinase is highly similar to the gene products of S. cerevisiae cdc28, and S. pombe cdc2. It is a catalytic subunit of the cyclin-dependent protein kinase complex, whose activity is restricted to the G1-S phase, and essential for cell cycle G1/S phase transition. This protein associates with and regulated by the regulatory subunits of the complex including cyclin A or E, CDK inhibitor p21Cip1 (CDKN1A) and p27Kip1 (CDKN1B). Its activity is also regulated by its protein phosphorylation. Two alternatively spliced variants and multiple transcription initiation sites of this gene have been reported. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | CDK2 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Descripción | Rabbit polyclonal antibody raised against synthetic phosphopeptide of CDK2. |
| Dilución | Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
| Formulación | In PBS (0.09% sodium azide) |
| génica | CDK2 |
| N.º de referencia del gen | NP_001789;P24941 |
| Alias de gen | p33(CDK2) |
| Mostrar más |
For Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.